Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLS3 Polyclonal Antibody
PLS3 Polyclonal Antibody
Supplier: Thermo Scientific PA567403
Description
PLS3 Polyclonal Antibody for ICC/IF

Specifications
PLS3 | |
Polyclonal | |
Unconjugated | |
PLS3 | |
AI115446; AL024105; BMND18; LOW QUALITY PROTEIN: plastin-3; MGC128570 protein; plastin 3; plastin 3 (T isoform); plastin 3 (T-isoform); plastin 3, T; plastin-3; PLS3; T fimbrin; T plastin; T-fimbrin; T-plastin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5358 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P13797 | |
PLS3 | |
Recombinant Human PLS3. Recombinant protein control fragment (Product #RP-108067). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction