Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLSCR1 Monoclonal Antibody (13A6 [TEC-23])
GREENER_CHOICE

Catalog No. PIMA528630
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA528630 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA528630 Supplier Thermo Scientific Supplier No. MA528630
Add to Cart
Add to Cart

PLSCR1 Monoclonal Antibody (13A6 [TEC-23])

PLSCR1 Monoclonal Antibody for Western Blot, ICC/IF, Flow, IP

TRUSTED_SUSTAINABILITY

Specifications

Antigen PLSCR1
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 13A6 [TEC-23]
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation Ascites with 15 mM sodium azide
Gene PLSCR1
Gene Accession No. P58195
Gene Alias ca(2+)-dependent phospholipid scramblase 1; Erythrocyte phospholipid scramblase; MmTRA1a; MmTRA1b; NOR1; Phospholipid scramblase 1; phospholipid scramblase 2; PL scramblase 1; Plscr1; Tra1; Tra1a; Tra1b; transplantability-associated protein 1; Tras1; Tras2
Gene Symbols PLSCR1
Host Species Mouse
Immunogen Pooled lipid rafts isolated from RBL-2H3 cells.
Purification Method Nan
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117540
Target Species Rat
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.