Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLSCR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104017 Supplier Invitrogen™ Supplier No. PA5104017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PLSCR1.

The calcium-dependent plasma membrane protein phospholipid scramblase 1 (PLSCR1) contributes to the transbilayer movement of phosphatidylserine and other membrane phospholipids upon influx of calcium into the cytosol. This movement results in plasma membrane phospholipid remodelling and surface exposure of phosphatidylserine in injured or apoptotic cells, which leads to cell death. Interferons and other cytokines induce expression of PLSCR1, implying that PLSCR1 also functions in cytokine signaling pathways. EGF stilmulation results in tyrosine phosphorylation of PLSCR1 on tyrosines 69 and 74, which allows it to interact with Shc, and thereby connecting Src kinase activation to stimulation of the EGF receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLSCR1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLSCR1
Gene Accession No. O15162, Q9JJ00
Gene Alias ca(2+)-dependent phospholipid scramblase 1; Erythrocyte phospholipid scramblase; MmTRA1a; MmTRA1b; NOR1; Phospholipid scramblase 1; phospholipid scramblase 2; PL scramblase 1; Plscr1; Tra1; Tra1a; Tra1b; transplantability-associated protein 1; Tras1; Tras2
Gene Symbols PLSCR1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLSCR1(Accession O15162), corresponding to amino acid residues T15-N65.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22038, 5359
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.