Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLSCR2 Monoclonal Antibody (OTI7A5)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527367
Description
PLSCR2 Monoclonal Antibody for Western Blot
May mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system.; FUNCTION: Isoform 1 has no prospholipid scramblase activity, due to the lack of a N-terminal proline-rich domain.
Specifications
PLSCR2 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9DCW2, Q9NRY7 | |
Plscr2 | |
Full length human recombinant protein of PLSCR2 produced in E.coli. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2b |
Western Blot | |
OTI7A5 | |
Unconjugated | |
Plscr2 | |
Ca(2+)-dependent phospholipid scramblase 2; phospholipid scramblase 2; PL scramblase 2; Plscr2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
18828, 57047 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction