Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLSCR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104018
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104018 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104018 Supplier Invitrogen™ Supplier No. PA5104018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PLSCR2.

May mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system.; FUNCTION: Isoform 1 has no prospholipid scramblase activity, due to the lack of a N-terminal proline-rich domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLSCR2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Plscr2
Gene Accession No. Q9DCW2, Q9NRY7
Gene Alias Ca(2+)-dependent phospholipid scramblase 2; phospholipid scramblase 2; PL scramblase 2; Plscr2
Gene Symbols Plscr2
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLSCR2(Accession Q9NRY7), corresponding to amino acid residues A185-I235.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18828, 57047
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.