Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLSCR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544769
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544769 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544769 Supplier Invitrogen™ Supplier No. PA544769
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GCGTDTNFEV KTRDESRSVG RISKQWGGLV REALTDADDF GLQFPLDLDV Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

May mediate accelerated ATP-independent bidirectional transbilayer migration of phospholipids upon binding calcium ions that results in a loss of phospholipid asymmetry in the plasma membrane. May play a central role in the initiation of fibrin clot formation, in the activation of mast cells and in the recognition of apoptotic and injured cells by the reticuloendothelial system. Seems to play a role in apoptosis, through translocation of cardiolipin from the inner to the outer mitochondrial membrane which promotes BID recruitment and enhances tBid-induced mitochondrial damages. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLSCR3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Plscr3
Gene Accession No. Q9NRY6
Gene Alias 2210403O21Rik; 2610037N06Rik; Ca 2+ dependent phospholipid scramblase 3; ca(2+)-dependent phospholipid scramblase 3; ESTM3; LOW QUALITY PROTEIN: phospholipid scramblase 3; LOW QUALITY PROTEIN: phospholipid scramblase 3; phospholipid scramblase 3; phospholipid scramblase 3; PL scramblase 3; Pls3; Plscr3; Unknown (protein for MGC:138060); X83310
Gene Symbols Plscr3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human PLSCR3.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57048
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.