Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLSCR4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104019
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104019 Supplier Thermo Scientific Supplier No. PA5104019
Add to Cart
Add to Cart

PLSCR4 Polyclonal Antibody

PLSCR4 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PLSCR4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLSCR4
Gene Accession No. Q9NRQ2
Gene Alias AV245873; Ca(2+)-dependent phospholipid scramblase 4; cell growth inhibiting protein 43; Cell growth-inhibiting gene 43 protein; GIG43; Phospholipid scramblase 4; PL scramblase 4; Pls4; PLSCR4; TRA1
Gene Symbols PLSCR4
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLSCR4(Accession Q9NRQ2), corresponding to amino acid residues D162-Q212.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57088
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.