Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLTP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 8 publications
$206.50 - $499.50
Specifications
Antigen | PLTP |
---|---|
Dilution | Western Blot 1:500, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:200 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB400106SS
![]() |
Novus Biologicals
NB400106SS |
0.05 mL |
Each for $206.50
|
|
|||||
NB400106
![]() |
Novus Biologicals
NB400106 |
0.2 mL |
Each for $499.50
|
|
|||||
Description
PLTP Polyclonal specifically detects PLTP in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
PLTP | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cancer, Cholesterol Metabolism, Diabetes Research, Lipid and Metabolism, Signal Transduction | |
Whole antisera with 0.02% Sodium Azide | |
HDLCQ9, Lipid transfer protein II, phospholipid transfer protein | |
PLTP | |
IgG | |
Unpurified | |
55 kDa |
Western Blot 1:500, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:200 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Mouse | |
P55058 | |
5360 | |
A partial peptide of human PLTP. [UniProt# P55058] | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title