Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PluriSTEM-XF™ Recombinant Vitronectin

Catalog No. cc130 Shop All MilliporeSigma Products
Click to view available options
Quantity:
500 μg

For Cell attachment, Stem cell culture, Cell culture applications

TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.5 mg/mL
For Use With (Application) Cell Attachment and Proliferation, Stem Cell Culture, Cell Culture
Quantity 500 μg
Product Type Recombinant Vitronectin
Content And Storage Store at -20°C until expiration date on vial. Once thawed, product is stable at 2° to 8°C for two weeks. Thawed vitronectin can be aliquoted and stored at -20°C. Avoid additional freeze-thaw cycles.
Purity or Quality Grade ≥95% by SDS-PAGE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.