Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

StatLab™ Plus Alcohol
GSA_VASDP

Catalog No. stl71001
Click to view available options
Quantity:
4 x 1 gal.
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.
This item is not returnable. View return policy
Harmful if swallowed, if in contact with skin, or if inhaled.

A custom isopropyl and methanol blend that can be used as a substitute for traditional ethanol blends.

  • 100% or 95% Plus alcohol (100% Plus alcohol with 5 parts deionized water)
  • Does not over-dehydrate small specimens
  • Great dehydrant for both small and large tissue specimens
  • May also be used in staining procedures

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Concentration 1
Quantity 4 x 1 gal.
Format Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.