Learn More
Description
Urea
- Provides highest resolution and the cleanest results for IEF performed under denaturing conditions
- Urea solubilizes and unfolds most proteins to their fully random conformation, with all ionizable groups exposed to solution
- 8M urea included in the 2-D sample solution
Low toxicity and environmentally safe solvent
- Used for covalently attach polyacrylamide gels to a glass surface
- Firmly attaches to the glass during staining and drying procedures
Urea:
- Molecular Weight: 60.06
- Physical state: Solid
- Color: White
- Pack of 500g
- Molecular Weight: 129.06
- Physical state: Liquid
- Color: Colorless
- Pack of 500mL
- Molecular Weight: 248.4
- Physical state: Liquid
- Color: Pale
- Pack of 25mL
IEF, PAGE, sequencing
Certifications
Suitable for DIGE
Specifications
Specifications
| CAS | 2530-85-0 |
| Description | PlusOne Repel-Silane ES |
| Color | Colorless |
| Concentration | 2% |
| Form | Liquid |
| For Use With (Application) | Used in IEF, PAGE and Sequencing |
| Molecular Weight (g/mol) | 129.06 |
| Quantity | 500 mL |
| Storage Requirements | Dry and dark at room temperature |
| Product Type | Urea |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.