Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLVAP Antibody, Novus Biologicals™
SDP

Catalog No. p-200048260 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP183911 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP183911 Supplier Novus Biologicals Supplier No. NBP183911
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

PLVAP Polyclonal specifically detects PLVAP in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Electron Microscopy.

Specifications

Antigen PLVAP
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoassay
Classification Polyclonal
Concentration 0.1mg/mL
Conjugate Unconjugated
Dilution Western Blot Validated by verified customer review., Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000, Electron Microscopy Reported in scientific literature (PMID:34280928).
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias FELSfenestrated-endothelial linked structure protein; PV-1 protein, Fenestrated endothelial-linked structure protein, gp68, plasmalemma vesicle associated protein, PV1Plasmalemma vesicle protein 1, PV-1plasmalemma vesicle-associated protein
Gene Symbols PLVAP
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:KEQLQKVQALCLPLDKDKFEMDLRNLWRDSIIPRSLDNLGYNLYHPLGSELASIRRACDHMPSLMSSKVEELARSLRADIERVARENSDLQRQKLEAQQGLRASQEAKQKVEKEAQAREAKLQAECSR
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83483
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.