Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLVAP Monoclonal Antibody (174/2)
GREENER_CHOICE

Catalog No. FERMA549086
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549086 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549086 Supplier Invitrogen™ Supplier No. MA549086
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PLVAP Monoclonal Antibody for Flow, ICC/IF, IHC (F), IP, Western Blot

PLVAP involved in the formation of stomatal and fenestral diaphragms of caveolae. This protein may function in microvascular permeability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLVAP
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 174/2
Concentration 200 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and 0.02% sodium azide
Gene PLVAP
Gene Accession No. Q9BX97
Gene Alias FELS; Fenestrated endothelial-linked structure protein; fenestrated-endothelial linked structure protein; PV-1 protein; gp68; MECA32; MECA-32 antigen; plasmalemma vesicle associated protein; plasmalemma vesicle protein 1; Plasmalemma vesicle-associated protein; PLVAP; Pv1; PV-1; PV-1 protein
Gene Symbols PLVAP
Host Species Mouse
Immunogen The monoclonal antibody 174/2 reacts with human plasmalemma vesicle associated protein (PLVAP).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 83483
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.