Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLVAP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115774
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115774 Supplier Invitrogen™ Supplier No. PA5115774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PLVAP.

PLVAP involved in the formation of stomatal and fenestral diaphragms of caveolae. This protein may function in microvascular permeability.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLVAP
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLVAP
Gene Accession No. Q91VC4, Q9BX97, Q9WV78
Gene Alias FELS; Fenestrated endothelial-linked structure protein; fenestrated-endothelial linked structure protein; PV-1 protein; gp68; MECA32; MECA-32 antigen; plasmalemma vesicle associated protein; plasmalemma vesicle protein 1; Plasmalemma vesicle-associated protein; PLVAP; Pv1; PV-1; PV-1 protein
Gene Symbols PLVAP
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLVAP(Accession Q9BX97), corresponding to amino acid residues I390-S440.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56765, 83483, 84094
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.