Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLXDC2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA520466
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520466 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520466 Supplier Thermo Scientific Supplier No. PA520466

Rabbit Polyclonal Antibody

A suggested positive control is human brain tissue lysate. PA5-20466 can be used with blocking peptide PEP-0583.

May play a role in tumor angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXDC2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene PLXDC2
Gene Accession No. Q6UX71, Q9DC11
Gene Alias 1200007L24Rik; 5430431D22Rik; AI646728; AU022916; AV231634; plexin domain containing 2; plexin domain-containing protein 2; PLXDC2; TEM7R; tumor endothelial marker 7 related; tumor endothelial marker 7-related protein; UNQ2514/PRO6003
Gene Symbols PLXDC2
Host Species Rabbit
Immunogen An 18 amino acid peptide from near the amino terminus of human Plxdc2.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361282, 67448, 84898
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.