Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLXNA3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547571
Description
In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Plexin A1, rmPlexin A2, recombinant human (rh) Plexin B1, rhPlexin C1, and rhPlexin D1 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
Coreceptor for SEMA3A and SEMA3F. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance in the developing nervous system. Regulates the migration of sympathetic neurons, but not of neural crest precursors. Required for normal dendrite spine morphology in pyramidal neurons. May play a role in regulating semaphorin-mediated programmed cell death in the developing nervous system. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.
Specifications
| PLXNA3 | |
| Polyclonal | |
| Unconjugated | |
| Plxna3 | |
| 6.3; HSSEXGENE; mKIAA4078; PlexA3; plexin 3; Plexin 4, SEX, homolog to the cMet/HGF receptors; plexin A3; plexin-4; Plexin-A3; Plxa3; Plxn3; Plxn4; Plxna3; Semaphorin receptor SEX; SEX; Sex chromosome X transmembrane protein of HGF receptor family 3; SEXPLEXIN-A3; XAP-6 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 18846, 309280 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| D3ZPX4, P70208 | |
| Plxna3 | |
| E. coli-derived recombinant mouse Plexin A3 Leu34-Asp150. | |
| 100 μg | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction