Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLXNA4 Monoclonal Antibody (F.566.2)
GREENER_CHOICE

Catalog No. PIMA514795
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA514795 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA514795 Supplier Invitrogen™ Supplier No. MA514795
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

It is not recommended to aliquot this antibody.

PLXNA4 is a protein coding gene. Coreceptor for SEMA3A. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance in the developing nervous system. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNA4
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone F.566.2
Concentration 155 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and <0.02% sodium azide; pH 7.5
Gene Plxna4
Gene Accession No. Q80UG2, Q9HCM2
Gene Alias 9330117B14; FAYV2820; Kiaa1550; mKIAA1550; OTTHUMP00000207245; PLEXA4; plexin A4; plexin A4, A; plexin-A4; Plxa4; Plxna4; Plxna4a; PLXNA4B; PRO34003; UNQ2820/PRO34003
Gene Symbols Plxna4
Host Species Rabbit
Immunogen GST-fusion protein corresponding to residues from the human sequence of Plexin A4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243743, 312213, 91584
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.