Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLXNB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143747
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143747 Supplier Invitrogen™ Supplier No. PA5143747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects 300, 150, and 100 kDa* bands on SDS-PAGE immunoblots of mouse liver and rat PC12 cells. These bands correspond to the full length and proteolytically processed forms of Plexin B1. In mouse liver, this antibody detects 300 and 100 kDa forms of Plexin B1.

Plexin B1 is a protein of the plexin family that in humans is encoded by the PLXNB1gene. Receptor for SEMA4D. Plays a role in RHOA activation and subsequent changes of the actin cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNB1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene Plxnb1
Gene Accession No. O43157, Q8CJH3
Gene Alias 2900002G15Rik; AU042020; Kiaa0407; mKIAA0407; plexin 1; plexin 5; plexin B1; plexin-A1; PLEXIN-B1; PLXN5; Plxnb1; Semaphorin receptor SEP; SEP
Gene Symbols Plxnb1
Host Species Rabbit
Immunogen A synthetic peptide (coupled to KLH) corresponding to amino acid residues in the C-terminal region of human Plexin B1. This sequence has high homology with similar regions in rat and mouse Plexin B1, and has low homology to other plexin family members.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235611, 316009, 5364
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.