Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLXNB1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA587560

Catalog No. PIPA587560


Only null left
Add to Cart

Description

Description

Immunogen sequence: RVCLRDQHYY SYVELPLACE GGRYGLIQAA AVATSREVAH GEVLFAAFSS AAPPTVGRPP SAAAGASGAS ALCAFPLDEV DRLANRTRDA CYTREGRAED GTEVAYIEYD VNSDCAQLPV DTLDAYPCGS DHTPSPMASR VPLEATPILE WPGIQLTAVA VTMEDGHTIA FLGDSQGQLH RVYLGPGSDG HPYSTQSIQQ GSAVSRDLTF DGTFEHLYVM T; Positive Samples: LO2, Mouse lung; Cellular Location: Cell membrane, Single-pass type I membrane protein.

Plexin B1 is a protein of the plexin family that in humans is encoded by the PLXNB1gene. Receptor for SEMA4D. Plays a role in RHOA activation and subsequent changes of the actin cytoskeleton. Plays a role in axon guidance, invasive growth and cell migration.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLXNB1
Polyclonal
Unconjugated
Plxnb1
2900002G15Rik; AU042020; Kiaa0407; mKIAA0407; plexin 1; plexin 5; plexin B1; plexin-A1; PLEXIN-B1; PLXN5; Plxnb1; Semaphorin receptor SEP; SEP
Rabbit
Affinity chromatography
RUO
235611, 5364
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot
0.47 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43157, Q8CJH3
Plxnb1
Recombinant fusion protein containing a sequence corresponding to amino acids 250-470 of human PLXNB1 (NP_0026642).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.