Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLXNB2 Monoclonal Antibody (537237)
GREENER_CHOICE

Catalog No. PIMA524237
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524237 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524237 Supplier Invitrogen™ Supplier No. MA524237
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

In direct ELISAs, no cross-reactivity with recombinant human Plexin A4, B1, B3, C1, D1, or recombinant mouse Plexin A1, A2, or A3 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Plexin-B2 is a member of the Semaphorin receptor family. The semaphorin family consists of membrane and secreted proteins involved in attraction and repulsion of neuronal guidance cues. Plexin-B1 is a transmembrane protein that can be cleaved converting the single chain precursor into a heterodimeric receptor. It has been shown to bind Sema4D/CD100 and is involved in axon development in mouse embryos. Sema4D also plays a role in the immune system interacting with CD72. Plexin-B2's possible role in the immune system has not been characterized.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNB2
Applications Western Blot
Classification Monoclonal
Clone 537237
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene PLXNB2
Gene Accession No. O15031
Gene Alias Debt; differentially expressed in brain tumors; dJ402G11.3; KIAA0315; MM1; MM1KIAA0315dJ402G11.3; Nbla00445; PLEXB2; plexin B2; plexin-B2; PLXNB2
Gene Symbols PLXNB2
Host Species Mouse
Immunogen CHO-derived recombinant human Plexin B2 Leu20-Arg1160.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23654
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.