Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLXNB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA548065
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA548065 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA548065 Supplier Invitrogen™ Supplier No. PA548065
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Plexin B3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the plexin family. It functions as a receptor for semaphorin 5A, and plays a role in axon guidance, invasive growth and cell migration. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNB3
Applications Flow Cytometry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Plxnb3
Gene Accession No. Q9ULL4
Gene Alias AI451018; Ensembl:ENSRNOP00000053235}; FLJ76953; KIAA1206; PLEXB3; plexin 6; plexin B3; plexin-6; Plexin-B3; plexin-B3 {ECO:0000303; PLEXR; Plxn6; PLXNB3; plxnb3 {ECO:0000312; Protein Plxnb3; PubMed:20696765}; RGD1560615
Gene Symbols Plxnb3
Host Species Sheep
Immunogen CHO-derived recombinant human Plexin B3 His45-Gln1255 (Glu1156Asp).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5365
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.