Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLXNB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590030
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590030 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590030 Supplier Thermo Scientific Supplier No. PA590030
Add to Cart
Add to Cart

PLXNB3 Polyclonal Antibody

PLXNB3 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNB3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.02 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Plxnb3
Gene Accession No. D3ZLH5
Gene Alias AI451018; Ensembl:ENSRNOP00000053235}; FLJ76953; KIAA1206; PLEXB3; plexin 6; plexin B3; plexin-6; Plexin-B3; plexin-B3 {ECO:0000303; PLEXR; Plxn6; PLXNB3; plxnb3 {ECO:0000312; Protein Plxnb3; PubMed:20696765}; RGD1560615
Gene Symbols Plxnb3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1050-1250 of human PLXNB3 (NP_0011567291).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363517
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.