Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLXNB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA545692
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA545692 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA545692 Supplier Invitrogen™ Supplier No. PA545692
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DYRTYAERAF FPGHGGCPLQ PKPEGPGEDG HCATVRQGLT QLSNLLNSKL Sequence homology: Guinea Pig: 83%; Human: 100%.

The protein encoded by this gene is a member of the plexin family. It functions as a receptor for semaphorin 5A, and plays a role in axon guidance, invasive growth and cell migration. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNB3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Plxnb3
Gene Accession No. Q9ULL4
Gene Alias AI451018; Ensembl:ENSRNOP00000053235}; FLJ76953; KIAA1206; PLEXB3; plexin 6; plexin B3; plexin-6; Plexin-B3; plexin-B3 {ECO:0000303; PLEXR; Plxn6; PLXNB3; plxnb3 {ECO:0000312; Protein Plxnb3; PubMed:20696765}; RGD1560615
Gene Symbols Plxnb3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human PLXNB3 (aa 1332-1381).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5365
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.