Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLXNC1 Monoclonal Antibody (2H11C11)
GREENER_CHOICE

Catalog No. FERMA548682
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548682 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548682 Supplier Invitrogen™ Supplier No. MA548682
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PLXNC1 Monoclonal Antibody for Western Blot, IHC, Flow, ELISA

Receptor for SEMA7A, for smallpox semaphorin A39R, vaccinia virus semaphorin A39R and for herpesvirus Sema protein. Binding of semaphorins triggers cellular responses leading to the rearrangement of the cytoskeleton and to secretion of IL6 and IL8. {ECO:0000250, ECO:0000269; PubMed:20727575}.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXNC1
Applications ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2H11C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PLXNC1
Gene Accession No. O60486
Gene Alias 2510048K12Rik; AW742158; CD232; CD232 antigen; plexin (semaphorin receptor); plexin C1; plexin-C1; PLXNC1; PLXN-C1; receptor for viral semaphorin protein; receptor for virally-encoded semaphorin; Vespr; VESPRplexin-C1; viral-encoded semaphorin protein receptor; virus-encoded semaphorin protein receptor
Gene Symbols PLXNC1
Host Species Mouse
Immunogen Purified recombinant fragment of human CD232 (AA: extra 35-234) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10154
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.