Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLXNC1 Monoclonal Antibody (3C10B2)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548683
Description
PLXNC1 Monoclonal Antibody for Western Blot, Flow, ELISA
Receptor for SEMA7A, for smallpox semaphorin A39R, vaccinia virus semaphorin A39R and for herpesvirus Sema protein. Binding of semaphorins triggers cellular responses leading to the rearrangement of the cytoskeleton and to secretion of IL6 and IL8. {ECO:0000250, ECO:0000269; PubMed:20727575}.
Specifications
PLXNC1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O60486 | |
PLXNC1 | |
Purified recombinant fragment of human CD232 (AA: extra 35-234) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
3C10B2 | |
Unconjugated | |
PLXNC1 | |
2510048K12Rik; AW742158; CD232; CD232 antigen; plexin (semaphorin receptor); plexin C1; plexin-C1; PLXNC1; PLXN-C1; receptor for viral semaphorin protein; receptor for virally-encoded semaphorin; Vespr; VESPRplexin-C1; viral-encoded semaphorin protein receptor; virus-encoded semaphorin protein receptor | |
Mouse | |
Protein G | |
RUO | |
10154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction