Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLXND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5143750
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5143750 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5143750 Supplier Invitrogen™ Supplier No. PA5143750
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects 215 and 195 kDa bands corresponding to the apparent molecular mass of Plexin D1 variants in human umbilical vein endothelial cells.

Cell surface receptor for SEMA4A and for class 3 semaphorins, such as SEMA3A, SEMA3C and SEMA3E. Plays an important role in cell-cell signaling, and in regulating the migration of a wide spectrum of cell types. Regulates the migration of thymocytes in the medulla. Regulates endothelial cell migration. Plays an important role in ensuring the specificity of synapse formation. Required for normal development of the heart and vasculature. Mediates anti-angiogenic signaling in response to SEMA3E. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLXND1
Applications Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide
Gene PLXND1
Gene Accession No. Q3UH93, Q9Y4D7
Gene Alias 6230425C21Rik; b2b1863Clo; b2b553Clo; KIAA0620; MGC75353; PLEXD1; plexin D1; plexin-D1; Plxnd1
Gene Symbols PLXND1
Host Species Rabbit
Immunogen Plexin D1 synthetic peptide (coupled to carrier protein) corresponding to amino acids from the C-terminal region of human Plexin D1. This sequence is well conserved in rat and mouse Plexin D1, and has some homology to other Plexin family members.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23129, 312652, 67784
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.