Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLZF Antibody (816421), Janelia Fluor™ 646, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB29441J
Description
PLZF Monoclonal antibody specifically detects PLZF in Mouse samples. It is validated for Flow Cytometry (Intracellular Staining)Specifications
PLZF | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry (Intracellular Staining) | |
816421 | |
Intracellular Staining by Flow Cytometry | |
PLZFzinc finger and BTB domain-containing protein 16, zinc finger and BTB domain containing 16, Zinc finger protein 145, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), Zinc finger protein PLZF, ZNF145Promyelocytic leukemia zinc finger protein | |
E. coli-derived recombinant mouse PLZF, Met1-Gln254, Accession # NP_001028496 | |
0.1 mL | |
Cancer, Innate Immunity, Stem Cell Markers | |
7704 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction