Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLZF Antibody (816421), mFluor Violet 610 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB29441MFV610
Description
PLZF Monoclonal antibody specifically detects PLZF in Mouse samples. It is validated for Flow CytometrySpecifications
| PLZF | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cancer, Innate Immunity, Stem Cell Markers | |
| 7704 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| 816421 | |
| Intracellular Staining by Flow Cytometry | |
| PLZFzinc finger and BTB domain-containing protein 16, zinc finger and BTB domain containing 16, Zinc finger protein 145, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), Zinc finger protein PLZF, ZNF145Promyelocytic leukemia zinc finger protein | |
| E. coli-derived recombinant mouse PLZF, Met1-Gln254, Accession # NP_001028496 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction