Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLZF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP219870
Description
PLZF Polyclonal antibody specifically detects PLZF in Human, Chicken samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen).Specifications
PLZF | |
Polyclonal | |
Unconjugated | |
PBS, 20% Glycerol (pH7). with 0.025% Proclin 300 | |
PLZFzinc finger and BTB domain-containing protein 16, zinc finger and BTB domain containing 16, Zinc finger protein 145, zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia), Zinc finger protein PLZF, ZNF145Promyelocytic leukemia zinc finger protein | |
Rabbit | |
74 kDa | |
0.1 mL | |
Cancer, Innate Immunity, Neuronal Stem Cell Markers, Stem Cell Markers | |
7704 | |
Human, Chicken | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
1mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:2000, Immunohistochemistry-Frozen | |
Q05516 | |
ZBTB16 | |
Recombinant protein encompassing a sequence within the center region of human PLZF. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Human. Species reactivity based on sequence identity: Mouse (97%), Rat (96%), Bovine (96%). | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction