Learn More
PLZF Monoclonal Antibody (Mags.21F7), Brilliant Violet™ 711, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$170.65 - $366.65
Specifications
Antigen | PLZF |
---|---|
Clone | Mags.21F7 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
40-793-2080
![]() |
Invitrogen™
407932080 |
25 μg |
Each
for $170.65
Save 12% Reg : |
|
|||||
40-793-2082
![]() ![]() |
Invitrogen™
407932082 |
100 μg |
Each
for $366.65
Save 8% Reg : |
|
|||||
Description
This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase. Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia (APL). Alternate transcriptional splice variants have been characterized.Specifications
PLZF | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with BSA and 0.09% sodium azide; pH 7.2 | |
AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); zinc finger protein PLZF; ZNF145 | |
ZBTB16 | |
Primary | |
4°C, store in dark, DO NOT FREEZE! |
Mags.21F7 | |
Flow Cytometry | |
Brilliant Violet 711 | |
Mouse | |
Human, Mouse | |
Q05516 | |
235320, 7704 | |
IgG1 κ | |
Affinity chromatography | |
ZBTB16 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.