Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLZF Monoclonal Antibody (Mags.21F7), PE, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody

$166.00 - $405.00

Specifications

Antigen PLZF
Clone Mags.21F7
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-112-2431
Encompass_Preferred
View Documents
Invitrogen™
12932080
25 μg
Each for $166.00
Only null left
Add to Cart
 
50-112-2432
Encompass_Preferred
View Documents
Invitrogen™
12932082
100 μg
Each for $405.00
Only null left
Add to Cart
 
Description

Description

Description: This Mags.21F7 monoclonal antibody reacts with human and mouse promyelocytic leukemia zinc finger (PLZF), a member of the BTB-POZ family of transcription factors. Expression of this transcriptional repressor in immune cells differs between mice and humans. In mice, PLZF is highly expressed in immature CD1d-resricted NKT2 and NKT1 cells, and a subset of gamma delta (Vg1.1+Vd6.3+) T cells. Studies have also demonstrated expression of PLZF in non-invariant CD1d-restricted T cells, as well as non-CD1d-restricted innate T cells. In humans, PLZF is expressed in NK cells, gamma delta T cells, as well as CD4 and CD8+ T cells. PLZF is also expressed in MR1-specific mucosal-associated invariant T cells, as well as in MHC Class II-restricted T cells that develop via a thymocyte-thymocyte interaction in humans. PLZF exists as a homodimer or in complex with PLZP, and has been shown to be involved in the development of NKT cells, NK cell function, cellular quiescence, and growth suppression. Finally, PLZF has been shown to inhibit gene expression induced by retinoic acid receptor. Applications Reported: This Mags.21F7 antibody has been reported for use in intracellular staining followed by flow cytometric analysis. Applications Tested: This Mags.21F7 antibody has been tested by intracellular staining followed by flow cytometric analysis of mouse lymph node cells using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol.

This gene is a member of the Krueppel C2H2-type zinc-finger protein family and encodes a zinc finger transcription factor that contains nine Kruppel-type zinc finger domains at the carboxyl terminus. This protein is located in the nucleus, is involved in cell cycle progression, and interacts with a histone deacetylase. Specific instances of aberrant gene rearrangement at this locus have been associated with acute promyelocytic leukemia (APL). Alternate transcriptional splice variants have been characterized.
Specifications

Specifications

PLZF
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
AI467657; Green's luxoid; lu; luxoid; Lx; PLZF; Polydactyly-luxate syndrome; promyelocytic leukaemia zinc finger; promyelocytic leukemia zinc finger; Promyelocytic leukemia zinc finger protein; ZBTB16; Zfp145; zinc finger and BTB domain containing 16; zinc finger and BTB domain containing 16 protein; zinc finger and BTB domain-containing protein 16; Zinc finger protein 145; zinc finger protein 145 (Kruppel-like, expressed in promyelocytic leukemia); zinc finger protein PLZF; ZNF145
ZBTB16
Primary
4°C, store in dark, DO NOT FREEZE!
ZBTB16
Mags.21F7
Flow Cytometry
PE
Mouse
Human, Mouse
Q05516
235320, 7704
IgG1 κ
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.