Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ EXOSC10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555873
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555873 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555873 Supplier Invitrogen™ Supplier No. PA555873
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FAHPYQYELN HFTPADAVLQ KPQPQLYRPI EETPCHFISS LDELVELNEK LLNCQEFAVD LEHHSYRSFL GLTCLMQIST RTEDF Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 89%.

About 50% of patients with polymyositis/scleroderma (PM-Scl) overlap syndrome are reported to have autoantibodies to a nuclear/nucleolar particle termed PM-Scl. Exosome component 10 (EXOSC10), also named autoantigen PM/Scl-2, is the 100 kDa antigen component of PM-Scl and is recognized by most sera of PM-Scl patients. EXOSC10 is strongly enriched in the nucleolus and a small amount has been found in cytoplasm supporting the existence of a nucleolar RNA exosome complex form. As a putative catalytic component ofthe RNA exosome complex which has 3'->5' exoribonuclease activity, EXOSC10 participates in a multitude of cellular RNA processing and degradation events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EXOSC10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene EXOSC10
Gene Accession No. Q01780
Gene Alias Autoantigen PM/Scl 2; Autoantigen PM/Scl 2 homolog; autoantigen PM-SCL; Exosc10; Exosome component 10; P100 polymyositis-scleroderma overlap syndrome-associated autoantigen; p2; p3; p4; PM/Scl-100; PMSCL; PM-Scl; PMSCL2; Polymyositis/scleroderma autoantigen 100 kDa; polymyositis/scleroderma autoantigen 2; Polymyositis/scleroderma autoantigen 2 homolog; RRP6; Rrp6p
Gene Symbols EXOSC10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PM-Scl. Recombinant protein control fragment (Product #RP-94455).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5394
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.