Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PM20D2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583139
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583139 Supplier Invitrogen™ Supplier No. PA583139
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AFTNLDVVFM AHPSQENAAY LPDMAEHDVT VKYYGKASHS ASYPWEGLNA LDAAVLAYNN LSVFRQQMKP TWR Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.

PM20D2 belongs to the peptidase M20A family. The function of PM20D2 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PM20D2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Pm20d2
Gene Accession No. Q8IYS1
Gene Alias β-alanyl-lysine dipeptidase; Acy1l2; aminoacylase 1-like 2; aminoacylase 1-like protein 2; aminoacylase-1-like protein 2; bA63L7.3; Gm424; peptidase M20 domain containing 2; peptidase M20 domain-containing protein 2; Pm20d2
Gene Symbols Pm20d2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PM20D2. Recombinant protein control fragment (Product #RP-104607).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 135293
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.