Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572853
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572853 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572853 Supplier Invitrogen™ Supplier No. PA572853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PMAT Polyclonal Antibody for Western Blot, IHC (P), ELISA

PMAT(Slc29a4) is a member of the SLC29 family and encodes a plasma membrane protein with 11 transmembrane helices. This protein catalyzes the re-uptake of monoamines into presynaptic neurons, thus determining the intensity and duration of monoamine neural signaling. It has been shown to transport several compounds, including serotonin, dopamine, and the neurotoxin 1-methyl-4-phenylpyridinium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMAT
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene SLC29A4
Gene Accession No. Q7RTT9
Gene Alias ENT4; Equilibrative nucleoside transporter 4; FLJ34923; hENT4; mPMAT; Plasma membrane monoamine transporter; PMAT; PSEC0113; SLC29A4; solute carrier family 29 (equilibrative nucleoside transporter), member 4; solute carrier family 29 (nucleoside transporters), member 4; solute carrier family 29 member 4
Gene Symbols SLC29A4
Host Species Rabbit
Immunogen SLC29A4 antibody for an 18 amino acid peptide near the center of human SLC29A4. The immunogen is within amino acids 310 - 360 of SLC29A4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 222962
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.