Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMCA ATPase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114932
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114932 Supplier Invitrogen™ Supplier No. PA5114932
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total Calcium Pump PMCA4 ATPase.

The calcium pump of the plasma membrane (PMCA ATPase) pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the sarcoplasmic and endoplasmic reticulum calcium (SERCA) ATPase and the sodium/potassium pump. Studies indicate that there are four known isoforms of the PMCA ATPase which are encoded by a multigene family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMCA ATPase
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene ATP2B1
Gene Accession No. G5E829, P11505, P11506, P20020, P23634, Q01814, Q16720, Q64542, Q64568, Q6Q477, Q9R0K7
Gene Alias 2810442I22Rik; 6430519O13Rik; ATP2B; ATP2B1; ATP2B2; ATP2B3; ATP2B4; ATPase isoform 2 Na+K+ transporting beta polypeptide 2; ATPase isoform 2, Na+K+ transporting, beta polypeptide 2; ATPase plasma membrane Ca2+ transporting 1; ATPase plasma membrane Ca2+ transporting 2; ATPase plasma membrane Ca2+ transporting 3; ATPase plasma membrane Ca2+ transporting 4; ATPase, Ca++ transporting, plasma membrane 1; ATPase, Ca++ transporting, plasma membrane 2; ATPase, Ca++ transporting, plasma membrane 3; ATPase, Ca++ transporting, plasma membrane 4; ATP-dependent plasma membrane calcium transporter; Ca2+/Mg2+ ATPase; CFAP39; cilia and flagella associated protein 39; CLA2; D6Abb2e; deaf waddler; dfw; E130111D10Rik; Gena300; jog; Matrix-remodeling-associated protein 1; MXRA1; OPCA; plasma membrane Ca(2+)-ATPase; plasma membrane Ca++-ATPase; plasma membrane Ca2+ pump 2; plasma membrane Ca2+-ATPases 4; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 1; plasma membrane calcium ATPase isoform 1b; Plasma membrane calcium ATPase isoform 2; Plasma membrane calcium ATPase isoform 3; Plasma membrane calcium ATPase isoform 4; plasma membrane calcium ATPase isoform 4a; plasma membrane calcium ATPase isoform 4b; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; plasma membrane calcium pump isoform 1b; Plasma membrane calcium pump isoform 2; Plasma membrane calcium pump isoform 3; Plasma membrane calcium pump isoform 4; Plasma membrane calcium-transporting ATPase 1; plasma membrane calcium-transporting ATPase 2; Plasma membrane calcium-transporting ATPase 3; plasma membrane calcium-transporting ATPase 4; plasma membrane calcium-transporting ATPase 4; plasma membrane calcium-transporting ATPase 1; PMCA; PMCA1; Pmca1a; Pmca1b; Pmca1c; PMCA1kb; PMCA2; PMCA2a; PMCA2i; Pmca3; PMCA3a; Pmca4; PMCA4a; PMCA4b; PMCA4x; sarcolemmal calcium pump; SCAX1; Tmy; wms; wri; wriggle
Gene Symbols ATP2B1, ATP2B2, ATP2B3, ATP2B4
Host Species Rabbit
Immunogen A synthesized peptide derived from human ATP2B4(Accession P23634), corresponding to amino acid residues K302-L352.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11941, 24215, 29598, 29599, 29600, 320707, 381290, 490, 491, 492, 493, 67972
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.