Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMCA1 ATPase Polyclonal Antibody, Invitrogen™

Catalog No. PA1914
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1914 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1914 Supplier Thermo Scientific Supplier No. PA1914
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA1-914 detects calcium pump of the plasma membrane 1a (PMCA1a ATPase) and 1b from rat, human and porcine tissues. PA1-914 has been successfully used in Western blot, immunohistochemistry (paraffin and frozen), and ELISA procedures. By Western blot, this antibody detects ∽134 kDa and ∽130 kDa proteins representing PMCA1a and 1b ATPase from rat, human and porcine samples. The PA1-914 immunogen is a synthetic peptide corresponding to the residues A(5) N N S V A Y S G V K N S I K E A N(22) of rat PMCA1 ATPase.

The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/potassium pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMCA1 ATPase
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene ATP2B1
Gene Accession No. P11505, P20020, P23220
Gene Alias 2810442I22Rik; Atp2b1; ATPase plasma membrane Ca2+ transporting 1; ATPase, Ca++ transporting, plasma membrane 1; E130111D10Rik; plasma membrane Ca2+ pump (PMCA1b); plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 1; plasma membrane calcium pump; Plasma membrane calcium pump isoform 1; Plasma membrane calcium-transporting ATPase 1; PMCA1; Pmca1a; Pmca1b; Pmca1c; PMCA1kb
Gene Symbols ATP2B1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the residues A(5) N N S V A Y S G V K N S I K E A N(22) of rat PMCA1 ATPase.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29598, 397636, 490
Target Species Human, Pig, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.