Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMCA2 ATPase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA593257
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA593257 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA593257 Supplier Invitrogen™ Supplier No. PA593257
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGDMTNSDFY SKNQRNESSH GGEFGCTMEE LRSLMELRGT EAVVKIKETY GDTEAICRRL KTSPVEGLPG TAPDLEKRKQ; Positive Samples: Mouse ovary, Mouse lung, Mouse liver, Rat liver; Cellular Location: Cell junction, Cell membrane, Multi-pass membrane protein, synapse.

The calcium pump of the plasma membrane, termed PMCA ATPase, pumps calcium from the cytosol to the extracellular space. This membrane-bound enzyme is related to a number of other ATPases including the SERCA ATPase and the sodium/K+ pump. There are four different genes encoding PMCA ATPase and studies have revealed 20 isoforms of the pump generated by alternate splicing of the primary gene products. mRNA distribution studies show that gene products 1 and 4 are transcribed in most tissues, however, products 2 and 3 are more tissue specific. Transcription of the splicing variants has also been found to be tissue specific. In the pancreas, where insulin secretion is calcium dependent, the beta cells only express the 4b isoform, however the alpha and gamma cells express both 4a and 4b isoforms. Studies have also shown that different splice variants have different affinities for calcium and calmodulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMCA2 ATPase
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ATP2B2
Gene Accession No. P11506, Q01814, Q9R0K7
Gene Alias Atp2b2; ATPase isoform 2 Na+K+ transporting beta polypeptide 2; ATPase isoform 2, Na+K+ transporting, beta polypeptide 2; ATPase plasma membrane Ca2+ transporting 2; ATPase, Ca++ transporting, plasma membrane 2; D6Abb2e; deaf waddler; dfw; Gena300; jog; plasma membrane Ca(2+)-ATPase; plasma membrane Ca++-ATPase; plasma membrane Ca2+ pump 2; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 2; plasma membrane calcium pump; Plasma membrane calcium pump isoform 2; Plasma membrane calcium-transporting ATPase 2; PMCA2; PMCA2a; PMCA2i; Tmy; wms; wri; wriggle
Gene Symbols ATP2B2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-80 of human ATP2B2 (NP_0016742).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11941, 24215, 491
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.