Learn More
Invitrogen™ PMCA3 ATPase Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA1916
Description
PA1-916 detects calcium pump of the plasma membrane 3 (PMCA3 ATPase) from rat, human and porcine tissues. PA1-916 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~127 kDa protein representing PMCA3a and PMCA3b ATPase from rat brain microsomal fractions. PA1-916 immunizing peptide corresponds to amino acid residues 5-22 from rat PMCA3 ATPase protein. This sequence is completely conserved between rat and human PMCA3 ATPase.
The protein encoded by this gene belongs to the family of P-type primary ion transport ATPases characterized by the formation of an aspartyl phosphate intermediate during the reaction cycle. These enzymes remove bivalent calcium ions from eukaryotic cells against very large concentration gradients and play a critical role in intracellular calcium homeostasis. The mammalian plasma membrane calcium ATPase isoforms are encoded by at least four separate genes and the diversity of these enzymes is further increased by alternative splicing of transcripts. The expression of different isoforms and splice variants is regulated in a developmental, tissue- and cell type-specific manner, suggesting that these pumps are functionally adapted to the physiological needs of particular cells and tissues. This gene encodes the plasma membrane calcium ATPase isoform 3. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
PMCA3 ATPase | |
Polyclonal | |
Unconjugated | |
ATP2B3 | |
6430519O13Rik; ATP2B3; ATPase plasma membrane Ca2+ transporting 3; ATPase, Ca++ transporting, plasma membrane 3; CFAP39; cilia and flagella associated protein 39; CLA2; LOW QUALITY PROTEIN: plasma membrane calcium-transporting ATPase 3; OPCA; plasma membrane calcium ATPase; Plasma membrane calcium ATPase isoform 3; plasma membrane calcium pump; Plasma membrane calcium pump isoform 3; Plasma membrane calcium-transporting ATPase 3; PMCA3; PMCA3a; SCAX1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100520061, 29599, 492 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Q16720, Q64568 | |
ATP2B3 | |
Synthetic Peptide: A(5) N S S I E F H P K P Q Q Q R E V P(22). | |
100 μg | |
Primary | |
Human, Rat, Pig | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.