Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMCA4b Mouse anti-Human, Chimpanzee, Unconjugated, Clone: JA3, MilliporeSigma™

Catalog No. MABN180125 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABN1801-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1801-25 Supplier MilliporeSigma Supplier No. MABN180125UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-PMCA4b, clone JA3, Cat. No. MABN1801, is a highly specific mouse monoclonal antibody that targets PMCA4b and has been tested in ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMCA4b
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone JA3
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Plasma membrane calcium-transporting ATPase 4;EC: 3.6.3.8;Matrix-remodeling-associated protein 1;Plasma membrane calcium ATPase isoform 4;Plasma membrane calcium pump isoform 4
Gene Symbols ATP2B4;ATP2B2;MXRA1
Host Species Mouse
Immunogen Purified plasma membrane calcium-transporting ATPase from human red blood cells.
Molecular Weight of Antigen ∽134 kDa observed; 137.92 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Test Specificity Clone JA3 detects Plasma membrane calcium-transporting ATPase in human brain tissue. It targets an epitope with in 25 amino acids from the C-terminal region.
Target Species Human, Chimpanzee
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.