Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PMCH Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583802
Description
Immunogen sequence: SIRNLDDDMV FNTFRLGKGF QKEDTAEKSV IAPSLEQYKN DESSFMNEEE NKVSKNTGSK HN.
MCH may act as a neurotransmitter or neuromodulator in a broad array of neuronal functions directed toward the regulation of goal-directed behavior, such as food intake, and general arousal. May also have a role in spermatocyte differentiation.
Specifications
PMCH | |
Polyclonal | |
Unconjugated | |
PMCH | |
A230109K23Rik; MCH; melanin-concentrating hormone; melanin-concentrating hormone precursor; NEI; Neuropeptide-glutamic acid-isoleucine; neuropeptide-glutamic acid-isoleucine (NEI); Neuropeptide-glycine-glutamic acid; neuropeptide-glycine-glutamic acid (NGE); NGE; Pmch; ppMCH; prepro-MCH; prepro-melanin-concentrating hormone; pro-MCH; pro-melanin concentrating hormone; pro-melanin-concentrating hormone | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110312, 5367 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P20382, P56942 | |
PMCH | |
Recombinant protein corresponding to Human PMCH. Recombinant protein control fragment (Product #RP-95718). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction