Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PME1 Mouse anti-Human, Mouse, Rat, Clone: 8A6-F8, MilliporeSigma™

Catalog No. MABC1183MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC1183MI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC1183MI Supplier MilliporeSigma Supplier No. MABC1183
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Protein phosphatase methylesterase 1 using this mouse monoclonal Anti-PME1 Antibody, clone 8A6-F8 Immunoprecipitation, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PME1
Applications Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 8A6-F8
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Ppme1,Pme1
Host Species Mouse
Immunogen His-tagged full length recombinant mouse Protein phosphatase methylesterase 1.
Purification Method Protein G Purified
Quantity 200 μL
Regulatory Status RUO
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Gene ID (Entrez) NP_082568.1
Test Specificity Clone 8A6-F8 specifically stains PME-1 in paraffin sections of human pancreas, testis, and cerebral cortex.
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.