Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMEL Monoclonal Antibody (1D4A5)
GREENER_CHOICE

Catalog No. FERMA548601
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548601 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548601 Supplier Invitrogen™ Supplier No. MA548601
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PMEL Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA

Melanoma is a malignant tumor of melanocytes which are found predominantly in skin but also in the bowel and the eye (see uveal melanoma). It is one of the rarer types of skin cancer but causes the majority of skin cancer related deaths.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMEL
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone 1D4A5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene PMEL
Gene Accession No. P40967
Gene Alias 95 kDa melanocyte-specific secreted glycoprotein; 95kDa melanocyte-specific secreted glycoprotein; D10H12S53E; D12S53E; D12S53Eh; gp100; gp87; M-alpha; M-beta; ME20; ME20M; ME20-M; ME20-S; Melanocyte lineage specific antigen GP100; melanocyte protein mel 17; melanocyte protein PMEL; Melanocyte protein Pmel 17; melanocytes lineage-specific antigen GP100; Melanoma-associated ME20 antigen; melanosomal matrix protein17; P1; P100; P26; PMEL; Pmel 17; PMEL17; premelanosome protein; Secreted melanoma-associated ME20 antigen; SI; SIL; SILV; silver homolog; silver locus protein; silver locus protein homolog; silver, mouse, homolog of
Gene Symbols PMEL
Host Species Mouse
Immunogen Purified recombinant fragment of human PMEL (AA: 25-192) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6490
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.