Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PMEL Monoclonal Antibody (5C6G8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548600
Description
PMEL Monoclonal Antibody for Western Blot, Flow, ELISA
Melanoma is a malignant tumor of melanocytes which are found predominantly in skin but also in the bowel and the eye (see uveal melanoma). It is one of the rarer types of skin cancer but causes the majority of skin cancer related deaths.
Specifications
PMEL | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P40967 | |
PMEL | |
Purified recombinant fragment of human PMEL (AA: 25-192) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Western Blot | |
5C6G8 | |
Unconjugated | |
PMEL | |
95 kDa melanocyte-specific secreted glycoprotein; 95kDa melanocyte-specific secreted glycoprotein; D10H12S53E; D12S53E; D12S53Eh; gp100; gp87; M-alpha; M-beta; ME20; ME20M; ME20-M; ME20-S; Melanocyte lineage specific antigen GP100; melanocyte protein mel 17; melanocyte protein PMEL; Melanocyte protein Pmel 17; melanocytes lineage-specific antigen GP100; Melanoma-associated ME20 antigen; melanosomal matrix protein17; P1; P100; P26; PMEL; Pmel 17; PMEL17; premelanosome protein; Secreted melanoma-associated ME20 antigen; SI; SIL; SILV; silver homolog; silver locus protein; silver locus protein homolog; silver, mouse, homolog of | |
Mouse | |
Protein G | |
RUO | |
6490 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction