Learn More
PMEL Mouse anti-Canine, Human, Rat, Clone: OTI8A4, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF500402

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Melanoma gp100 a melanosomal matrix protein whose expression is closely correlated with cellular melanin content and which is a frequent melanoma tumor antigen recognized by cytotoxic T lymphocytes. It is a likely candidate gene for some cases of human oculocutaneous albinism not associated with known albinism loci.Specifications
PMEL | |
Monoclonal | |
Unconjugated | |
PMEL | |
D12S53E, ME20, ME20-M, ME20M, P1, P100, PMEL17, SI, SIL, SILV, gp100 | |
Mouse | |
Affinity Chromatography | |
RUO | |
362818, 481102, 6490 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI8A4 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P40967 | |
PMEL | |
Full-length protein expressed in 293T cell transfected with human SILV expression vector | |
100 μg | |
Primary | |
Canine, Human, Rat | |
Antibody | |
IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.