Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMEL17/SILV Antibody (HMB45) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$534.50
Specifications
Antigen | PMEL17/SILV |
---|---|
Clone | HMB45 |
Concentration | 1.0 mg/mL |
Dilution | Western Blot 0.5-1.0ug/mL, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1ug/mL, Immunohistochemistry-Paraffin 0.5-1.0ug/mL, Flow (Intracellular), CyTOF-ready, Multiplex Immunoassay |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining), CyTOF, Multiplex |
Description
PMEL17/SILV Monoclonal antibody specifically detects PMEL17/SILV in Human, Canine (Negative), Rat (Negative) samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular), CyTOF, Multiplex ImmunoassaySpecifications
PMEL17/SILV | |
1.0 mg/mL | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining), CyTOF, Multiplex | |
Unconjugated | |
Mouse | |
Human, Canine (Negative), Rat (Negative) | |
D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
Extract of pigmented melanoma metastases from lymph nodes | |
Primary | |
Store at -20 to -80C. Avoid freeze-thaw cycles. |
HMB45 | |
Western Blot 0.5-1.0ug/mL, Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 0.5-1ug/mL, Immunohistochemistry-Paraffin 0.5-1.0ug/mL, Flow (Intracellular), CyTOF-ready, Multiplex Immunoassay | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS | |
6490 | |
IgG1 κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title