Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PMEL17/SILV Antibody (HMB45 + PMEL/783), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$246.61 - $542.38

Specifications

Antigen PMEL17/SILV
Clone HMB45 + PMEL/783
Concentration 0.2mg/mL
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24452300
SDP
View Documents
Novus Biologicals
NBP2445230.02MG
0.02 mg
Each for $246.61
Only null left
Add to Cart
 
NBP24452301
SDP
View Documents
Novus Biologicals
NBP2445230.1MG
0.1 mg
Each for $542.38
Only null left
Add to Cart
 
NBP24452302
SDP
View Documents
Novus Biologicals
NBP2445230.2MG
0.2 mg N/A N/A N/A
Description

Description

Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

PMEL17/SILV Monoclonal specifically detects PMEL17/SILV in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

PMEL17/SILV
0.2mg/mL
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Unconjugated
Mouse
Human
P40967
6490
Extract of pigmented melanoma metastases from lymph nodes (HMB45); Recombinant human SILV protein (PMEL/783)
Primary
Store at 4C.
95 kDa
HMB45 + PMEL/783
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 0.5 - 1.0 ug/ml
Monoclonal
Purified
RUO
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17
PMEL
IgG
Protein A or G purified
By immunohistochemistry, it specifically recognizes a protein in melanocytes and melanomas. This MAb reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. Metastatic amelanotic melanoma can often be confused with a variety of poorly differentiated carcinomas, large cell lymphomas, and sarcomas using H & E stains alone. It is also difficult to differentiate melanoma from spindle cell carcinomas and various types of mesenchymal neoplasms. This MAb stains fetal and neonatal melanocytes, junctional and blue nevus cells, and malignant melanoma. This MAb also stains Angiomyolipoma (PEComa).
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.