Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMEL17/SILV Antibody (NKI-beteb), Novus Biologicals™

Mouse Monoclonal Antibody
$524.00
Specifications
Antigen | PMEL17/SILV |
---|---|
Clone | NKI-beteb |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array, Flow (Intracellular) |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array, Flow Cytometry, Immunohistochemistry (Frozen) |
Description
PMEL17/SILV Monoclonal specifically detects PMEL17/SILV in Human, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array, Flow (Intracellular).Specifications
PMEL17/SILV | |
0.2 mg/ml | |
Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array, Flow Cytometry, Immunohistochemistry (Frozen) | |
Unconjugated | |
Mouse | |
Human, Equine | |
D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
PMEL | |
IgG2b κ | |
Protein A or G purified | |
By immunohistochemistry, it specifically recognizes a protein in melanocytes and melanomas. This monoclonal antibody reacts with junctional and blue nevus cells and variably with fetal and neonatal melanocytes. Intradermal nevi, normal adult melanocytes, and non-melanocytic cells are negative. It does not stain tumor cells of epithelial, lymphoid, glial, or mesenchymal origin. This Mab labels formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation. |
NKI-beteb | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array, Flow (Intracellular) | |
Monoclonal | |
Purified | |
RUO | |
P40967 | |
6490 | |
Lymph node pigmented melanoma metastases extract was used as the immunogen for the gp100 NKI-beteb antibody | |
Primary | |
Store at 4C. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title