Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMEL17/SILV Antibody (SPM142), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234739MFV450
Description
PMEL17/SILV Monoclonal antibody specifically detects PMEL17/SILV in Human, Canine (Negative), Rat (Negative) samples. It is validated for Flow CytometrySpecifications
PMEL17/SILV | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
6490 | |
Store at 4C in the dark. | |
IgG1 κ |
Flow Cytometry | |
SPM142 | |
Flow Cytometry | |
D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
Extract of pigmented melanoma metastases from lymph nodes | |
0.1 mL | |
Primary | |
Human, Canine (Negative), Rat (Negative) | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction