Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PMEL17/SILV Antibody (SR1021), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321629100UL
Description
PMEL17/SILV Monoclonal antibody specifically detects PMEL17/SILV in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
PMEL17/SILV | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
6490 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry | |
SR1021 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
D12S53EP1, gp100, ME20, ME20-M, melanocyte protein mel 17, Melanocyte protein Pmel 17, Melanocytes lineage-specific antigen GP100, Melanoma-associated ME20 antigen, melanosomal matrix protein17, PMEL17P100, premelanosome proteinME20M, SI, SIL, silver (mouse homolog) like, silver homolog (mouse), Silver locus protein homolog, silver, mouse, homolog of, SILVPmel17 | |
A synthesized peptide derived from human PMEL17/SILV (Uniprot #: P40967) | |
100 μg | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction