Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PMEPA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584894
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584894 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584894 Supplier Invitrogen™ Supplier No. PA584894
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VNSTAAAAAG QPNVSCTCNC KRSLFQSMEI TELE Highest antigen sequence indentity to the following orthologs: Rat - 86%, Mouse - 88%.

This gene encodes a transmembrane protein that contains a Smad interacting motif (SIM). Expression of this gene is induced by androgens and transforming growth factor beta, and the encoded protein suppresses the androgen receptor and transforming growth factor beta signaling pathways though interactions with Smad proteins. Overexpression of this gene may play a role in multiple types of cancer. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PMEPA1
Applications Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PMEPA1
Gene Accession No. Q969W9
Gene Alias 2210418I02Rik; AW455466; Erg1.2; N4wbp4; Nedd4 WW binding protein 4; NEDD4 WW domain-binding protein 4; PMEPA1; Prostate transmembrane protein androgen induced 1; prostate transmembrane protein, androgen induced 1; protein TMEPAI; Solid tumor-associated 1 protein; STAG1; Tmepai; transmembrane prostate androgen-induced protein; transmembrane, prostate androgen induced RNA
Gene Symbols PMEPA1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PMEPA1. Recombinant protein control fragment (Product #RP-105560).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56937
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.